Piperidin-3-ol 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L01333-04
L01333-09
L01333-18
CAAAAL01333-04EA
32.23
CAD
CAAAAL01333-04
CAAAAL01333-09
CAAAAL01333-18
Piperidin-3-ol 98+%
Piperidin-3-ol
Purity: 98+%
CAS No.: 6859-99-0
Molecular Formula: C5H11NO
Formula Weight: 101.15
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₅H₁₁NO Boiling Pt: 67…69 °C/2 mm Melting Pt: 56…60 °C Flash Pt: >100 °C (212 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014591 CAS Number: 6859-99-0 EINECS: 229-957-4 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...