2-Methylbiphenyl 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01312-03
L01312-06
CAAAAL01312-03EA
84.21
CAD
CAAAAL01312-03
CAAAAL01312-06
2-Methylbiphenyl 98%
2-Methylbiphenyl
Purity: 98%
CAS No.: 643-58-3
Molecular Formula: C13H12
Formula Weight: 168.24
Flash Point: 107°(224°F)
Refractive Index: 1.5910
Melting Point: 0°
Boiling Point: 256°
Beilstein Reference: 2039655
MDL No.: MFCD00008517
Formula:
C₁₃H₁₂ Boiling Pt: 256 °C Melting Pt: 0 °C Density: 1,010 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008517 CAS Number: 643-58-3 EINECS: 211-400-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...