3-(3-Methoxyphenyl)propionic acid 98+%
Supplier: Thermo Scientific Chemicals
L01304-06
L01304-14
CAAAAL01304-06EA
74.58
CAD
CAAAAL01304-06
CAAAAL01304-14
3-(3-Methoxyphenyl)propionic acid 98+%
3-(3-Methoxyphenyl)propionic acid
Purity: 98+%
CAS No.: 10516-71-9
Molecular Formula: C10H12O3
Formula Weight: 180.20
Melting Point: 43-47°
Beilstein Reference: 1874422
Formula:
C₁₀H₁₂O₃ Melting Pt: 43…47 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014027 CAS Number: 10516-71-9 EINECS: 234-049-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...