Tetrafluoro-p-benzoquinone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetrafluoroquinone, 2,3,5,6-Tetrafluoro-1,4-benzoquinone, Tetrafluoro-1,4-benzoquinone, p-Fluoranil, 4-Fluoranil
L01287-03
L01287-06
CAAAAL01287-03EA
86.74
CAD
CAAAAL01287-03
CAAAAL01287-06
Tetrafluoro-p-benzoquinone 97%
Tetrafluoro-p-benzoquinone
Purity: 97%
CAS No.: 527-21-9
Molecular Formula: C6F4O2
Formula Weight: 180.06
Melting Point: ca 184° subl.
Beilstein Reference: 1875039
Formula:
C₆F₄O₂ MW: 180.06 g/mol Boiling Pt: 133 Melting Pt: ∼ 184 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00001592 CAS Number: 527-21-9 EINECS: 208-411-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...