3-(3-Hydroxyphenyl)propionic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(3-Hydroxyphenyl)propanoic acid
L01279-06
L01279-14
CAAAAL01279-06EA
162.74
CAD
CAAAAL01279-06
CAAAAL01279-14
3-(3-Hydroxyphenyl)propionic acid 98+%
3-(3-Hydroxyphenyl)propionic acid
Purity: 98+%
CAS No.: 621-54-5
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 110-114°
Beilstein Reference: 1947445
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 110…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016554 CAS Number: 621-54-5 EINECS: 210-692-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...