9-Acetylanthracene 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-(9-Anthracenyl)acetone, 9-Anthryl methyl ketone, 1-(9-Anthracenyl)ethanone

L01278-06 L01278-14
CAAAAL01278-06EA 68.73 CAD
CAAAAL01278-06 CAAAAL01278-14
9-Acetylanthracene 96%
9-Acetylanthracene

Purity: 96% CAS No.: 784-04-3 Molecular Formula: C16H12O Formula Weight: 220.27 Melting Point: 72-76° Beilstein Reference: 1370056

Formula: C₁₆H₁₂O
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 72…76 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001259
CAS Number: 784-04-3
EINECS: 212-315-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR