4-Methoxybiphenyl 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Biphenyl-4-yl methyl ether, 4-Phenylanisole

L01260-06 L01260-14
CAAAAL01260-06EA 49.3 CAD
CAAAAL01260-06 CAAAAL01260-14
4-Methoxybiphenyl 98+%
4-Methoxybiphenyl

Purity: 98+% CAS No.: 613-37-6 Molecular Formula: C13H12O Formula Weight: 184.24 Melting Point: 86-90° Boiling Point: 157°/10mm MDL No.: MFCD00014897

Formula: C₁₃H₁₂O
MW: 184.24 g/mol
Boiling Pt: 157 °C/10 mm
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00014897
CAS Number: 613-37-6
EINECS: 210-339-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR