1-Dodecanesulfonyl chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

L01224-06 L01224-14
CAAAAL01224-06EA 134.62 CAD
CAAAAL01224-06 CAAAAL01224-14
1-Dodecanesulfonyl chloride 98+%
1-Dodecanesulphonyl chloride

Purity: 98+% CAS No.: 10147-40-7 Molecular Formula: C12H25ClO2S Formula Weight: 268.84 Melting Point: 39-42° Boiling Point: 126-130°/4mm UN No.: 3261 MDL No.: MFCD00014728

Formula: C₁₂H₂₅ClO₂S
Boiling Pt: 126…130 °C/4 mm
Melting Pt: 39…42 °C
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014728
CAS Number: 10147-40-7
EINECS: 233-416-8
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR