3-Iodoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01159-09
L01159-18
CAAAAL01159-09EA
100.01
CAD
CAAAAL01159-09
CAAAAL01159-18
3-Iodoaniline 98%
3-Iodoaniline
Purity: 98%
CAS No.: 626-01-7
Molecular Formula: C6H6IN
Formula Weight: 219.03
Flash Point:
Formula:
C₆H₆IN Boiling Pt: 145…146 °C/15 mm Melting Pt: 21…24 °C Density: 1.821 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007781 CAS Number: 626-01-7 EINECS: 210-922-7 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...