2,4-Nonanedione 98+%
Supplier: Thermo Scientific Chemicals
L01151-06
L01151-14
CAAAAL01151-06EA
156.1
CAD
CAAAAL01151-06
CAAAAL01151-14
2,4-Nonanedione 98+%
2,4-Nonanedione
Purity: 98+%
CAS No.: 6175-23-1
Molecular Formula: C9H16O2
Formula Weight: 156.23
Refractive Index: 1.4608
Boiling Point: 100-103°/19mm
MDL No.: MFCD00015276
Formula:
C₉H₁₆O₂ MW: 156.22 g/mol Boiling Pt: 100…103 °C (19 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00015276 CAS Number: 6175-23-1 EINECS: 228-219-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...