3-Methylacetylacetone mixture of tautomers 95%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01135-06
L01135-14
CAAAAL01135-06EA
60.99
CAD
CAAAAL01135-06
CAAAAL01135-14
3-Methylacetylacetone mixture of tautomers 95%, Technical Grade
3-Methylacetylacetone
Purity: 95%
CAS No.: 815-57-6
Molecular Formula: C6H10O2
Formula Weight: 114.14
Flash Point: 56°(132°F)
Refractive Index: 1.4420
Boiling Point: 170-171°
Beilstein Reference: 635909
UN No.: 1224
MDL No.: MFCD00008762
Formula:
C₆H₁₀O₂ Boiling Pt: 170…171 °C Melting Pt: –31 °C Density: 0.980 Flash Pt: 56 °C(132 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008762 CAS Number: 815-57-6 EINECS: 212-420-3 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...