(Bromomethyl)cyclopropane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L01122-03
L01122-06
L01122-14
CAAAAL01122-03EA
73.31
CAD
CAAAAL01122-03
CAAAAL01122-06
CAAAAL01122-14
(Bromomethyl)cyclopropane 97%
(Bromomethyl)cyclopropane
Purity: 97%
CAS No.: 7051-34-5
Molecular Formula: C4H7Br
Formula Weight: 135.00
Storage Temperature: 2°C to 8°C
Flash Point: 41°(105°F)
Refractive Index: 1.4770
Boiling Point: 111-112°
Beilstein Reference: 605296
UN No.: 1993
MDL No.: MFCD00001306
Formula:
C₄H₇Br Boiling Pt: 111…112 °C Density: 1.410 Flash Pt: 41 °C (105 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001306 CAS Number: 7051-34-5 EINECS: 230-331-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...