(3-Methoxyphenoxy)acetic acid 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(3-Methoxyphenoxy)acetic acid
L01100-06
L01100-14
CAAAAL01100-06EA
67.31
CAD
CAAAAL01100-06
CAAAAL01100-14
(3-Methoxyphenoxy)acetic acid 97+%
(3-Methoxyphenoxy)acetic acid
Purity: 97+%
CAS No.: 2088-24-6
Molecular Formula: C9H10O4
Formula Weight: 182.18
Melting Point: 116-119°
Beilstein Reference: 1876898
Formula:
(C₆H₄(OCH₃))OCH₂COOH MW: 182.18 Melting Pt: 116…119 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014357 CAS Number: 2088-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...