3-Chlorobenzhydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Chlorobenzoic hydrazide
L01071-09
L01071-18
CAAAAL01071-09EA
148.52
CAD
CAAAAL01071-09
CAAAAL01071-18
3-Chlorobenzhydrazide 98+%
3-Chlorobenzhydrazide
Purity: 98+%
CAS No.: 1673-47-8
Molecular Formula: C7H7ClN2O
Formula Weight: 170.60
Melting Point: 156-160°
Beilstein Reference: 973243
Formula:
C₇H₇ClN₂O Melting Pt: 156…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014757 CAS Number: 1673-47-8 EINECS: 216-812-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...