Ethyl-3-chlorobenzoate 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Chlorobenzoic acid ethyl ester, Ethyl 3-chlorobenzoate

L01065-06 L01065-14
CAAAAL01065-06EA 34.92 CAD
CAAAAL01065-06 CAAAAL01065-14
Ethyl-3-chlorobenzoate 98+%
Ethyl 3-chlorobenzoate

Purity: 98+% CAS No.: 1128-76-3 Molecular Formula: C9H9ClO2 Formula Weight: 184.62 Refractive Index: 1.5210 Boiling Point: 118-120°/13mm MDL No.: MFCD00013634

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 118…120 °C/13 mm
Density: 1,186
Storage Temperature: Ambient
MDL Number: MFCD00013634
CAS Number: 1128-76-3
EINECS: 214-441-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR