4-Hydroxy-3,5-diiodobenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,5-Diiodo4-hydroxybenzaldehyde, 3,5-Diiodo-4-hydroxybenzaldehyde

L01060-09 L01060-18
CAAAAL01060-09EA 130.67 CAD
CAAAAL01060-09 CAAAAL01060-18
4-Hydroxy-3,5-diiodobenzaldehyde 98+%
4-Hydroxy-3,5-diiodobenzaldehyde

Purity: 98+% CAS No.: 1948-40-9 Molecular Formula: C7H4I2O2 Formula Weight: 373.92 Melting Point: 198-206° Beilstein Reference: 1283414

Formula: C₇H₄I₂O₂
Melting Pt: 198…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00014670
CAS Number: 1948-40-9
EINECS: 217-754-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR