2-Iodo-p-xylene 98+%

Supplier: Thermo Scientific Chemicals
Warning

L01055-09 L01055-18
CAAAAL01055-09EA 158 CAD
CAAAAL01055-09 CAAAAL01055-18
2-Iodo-p-xylene 98+%
2-Iodo-p-xylene

Purity: 98+% CAS No.: 1122-42-5 Molecular Formula: C8H9I Formula Weight: 232.07 Flash Point: 110°(230°F) Refractive Index: 1.5980 Boiling Point: 229-230° Beilstein Reference: 1905728 MDL No.: MFCD00013708

Formula: C₈H₉I
Boiling Pt: 229…230 °C
Density: 1,617
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013708
CAS Number: 1122-42-5
EINECS: 214-347-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR