(2-Methoxyethoxy)methyl chloride 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L01050-06
L01050-14
CAAAAL01050-06EA
79.16
CAD
CAAAAL01050-06
CAAAAL01050-14
(2-Methoxyethoxy)methyl chloride 94%
(2-Methoxyethoxy)methyl chloride
Purity: 94%
CAS No.: 3970-21-6
Molecular Formula: C4H9ClO2
Formula Weight: 124.57
Storage Temperature: -30°C to -10°C
Flash Point: 54°(129°F)
Refractive Index: 1.4270
Boiling Point: 50-52°/13mm
Beilstein Reference: 1900576
UN No.: 1992
MDL No.: MFCD00000888
Formula:
ClCH₂OCH₂CH₂OCH₃ Boiling Pt: 50…52 °C/13 mm Density: 1.094 Flash Pt: 54 °C (129 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00000888 CAS Number: 3970-21-6 EINECS: 223-589-8 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...