3-Methylbiphenyl 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Phenyltoluene

L01006-03 L01006-06
CAAAAL01006-03EA 91.64 CAD
CAAAAL01006-03 CAAAAL01006-06
3-Methylbiphenyl 95%
3-Methylbiphenyl

Purity: 95% CAS No.: 643-93-6 Molecular Formula: C13H12 Formula Weight: 168.24 Flash Point: 110°(230°F) Refractive Index: 1.6020 Melting Point: 4-5° Boiling Point: 272° Beilstein Reference: 2039658 MDL No.: MFCD00008533

Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 272 °C
Melting Pt: 4…5 °C
Density: 1.014
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008533
CAS Number: 643-93-6
EINECS: 211-404-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR