2-Bromobenzhydrazide 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromobenzene-1-carbohydrazide, 2-Bromobenzohydrazide, 2-Bromobenzoic acid hydrazide, 2-Bromobenzoic hydrazide
L00937-06
L00937-14
CAAAAL00937-06EA
66.04
CAD
CAAAAL00937-06
CAAAAL00937-14
2-Bromobenzhydrazide 98+%
2-Bromobenzhydrazide
Purity: 98+%
CAS No.: 29418-67-5
Molecular Formula: C7H7BrN2O
Formula Weight: 215.05
Melting Point: 152-154°
Beilstein Reference: 1941021
Formula:
C₇H₇BrN₂O Melting Pt: 152…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051577 CAS Number: 29418-67-5 EINECS: 249-614-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...