(2-Chlorophenoxy)acetic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00926-14
L00926-22
CAAAAL00926-14EA
131.14
CAD
CAAAAL00926-14
CAAAAL00926-22
(2-Chlorophenoxy)acetic acid 98+%
(2-Chlorophenoxy)acetic acid
Purity: 98+%
CAS No.: 614-61-9
Molecular Formula: C8H7ClO3
Formula Weight: 186.59
Melting Point: 147-152°
Beilstein Reference: 1103151
Formula:
(C₆H₄Cl)OCH₂COOH Boiling Pt: 308 °C (1013 hPa) Melting Pt: 147…152 °C Density: 1.509 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004298 CAS Number: 614-61-9 EINECS: 210-388-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...