o-Acetoxybenzoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

L00919-14 L00919-22
CAAAAL00919-14EA 237 CAD
CAAAAL00919-14 CAAAAL00919-22
o-Acetoxybenzoyl chloride 97%
o-Acetoxybenzoyl chloride

Purity: 97% CAS No.: 5538-51-2 Molecular Formula: C9H7ClO3 Formula Weight: 198.61 Flash Point: 110°(230°F) Odor: Pungent, makes eyes water Boiling Point: 134-136°/12mm Beilstein Reference: 880372 UN No.: 3261 MDL No.: MFCD00000663

Formula: C₉H₇ClO₃
Boiling Pt: 134…136 °C/12 mm
Melting Pt: 47…50 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000663
CAS Number: 5538-51-2
EINECS: 226-899-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR