Ethyl isothiocyanatoformate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00911-06
L00911-14
CAAAAL00911-06EA
117.24
CAD
CAAAAL00911-06
CAAAAL00911-14
Ethyl isothiocyanatoformate 97%
Ethyl isothiocyanatoformate
Purity: 97%
CAS No.: 16182-04-0
Molecular Formula: C4H5NO2S
Formula Weight: 131.15
Storage Temperature: 2°C to 8°C
Flash Point: 50°(122°F)
Refractive Index: 1.5030
Odor: Pungent
Boiling Point: 44-46°/10mm
Beilstein Reference: 606091
UN No.: 2924
MDL No.: MFCD00004814
Formula:
C₄H₅NO₂S Boiling Pt: 44…46 °C/10 mm Density: 1.110 Flash Pt: 50 °C (122 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004814 CAS Number: 16182-04-0 EINECS: 240-318-9 UN: 2929 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...