5-Hydroxyindole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Indol-5-ol, 5-Indolol, Indol-5-ol, 5-Hydroxy-1H-indole
L00858-03
L00858-06
CAAAAL00858-03EA
94.48
CAD
CAAAAL00858-03
CAAAAL00858-06
5-Hydroxyindole 98+%
5-Hydroxyindole
Purity: 98+%
CAS No.: 1953-54-4
Molecular Formula: C8H7NO
Formula Weight: 133.15
Storage Temperature: 2°C to 8°C
Melting Point: 106-111°
Beilstein Reference: 112349
MDL No.: MFCD00005677
Formula:
C₈H₇NO MW: 133.15 g/mol Melting Pt: 106…111 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005677 CAS Number: 1953-54-4 EINECS: 217-782-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...