2,6-Dinitrotoluene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00856-06
L00856-14
CAAAAL00856-06EA
46.93
CAD
CAAAAL00856-06
CAAAAL00856-14
2,6-Dinitrotoluene 97%
2,6-Dinitrotoluene
Purity: 97%
CAS No.: 606-20-2
Molecular Formula: C7H6N2O4
Formula Weight: 182.14
Flash Point: 207°(404°F)
Melting Point: 58-65°
Boiling Point: 285°
Beilstein Reference: 2052046
UN No.: 3454
MDL No.: MFCD00007158
Formula:
C₇H₆N₂O₄ MW: 182.14 g/mol Boiling Pt: 285 °C Melting Pt: 58…65 °C Density: 1.538 g/cm³ (15 °C) Flash Pt: 207 °C (404 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007158 CAS Number: 606-20-2 EINECS: 210-106-0 UN: 3454 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...