Trichloroacetic anhydride 95%

Supplier: Thermo Scientific Chemicals
Danger

L00849-18 L00849-30
CAAAAL00849-18EA 151.84 CAD
CAAAAL00849-18 CAAAAL00849-30
Trichloroacetic anhydride 95%
Trichloroacetic anhydride

Purity: 95% CAS No.: 4124-31-6 Molecular Formula: (Cl3CCO)2O Formula Weight: 308.76 Refractive Index: 1.4840 Boiling Point: 98-100°/11mm UN No.: 3265 MDL No.: MFCD00000793

Formula: C₄Cl₆O₃
MW: 308.76 g/mol
Boiling Pt: 98…100 °C (11 mmHg)
Melting Pt: -2
Density: 1.690
Storage Temperature: Ambient
MDL Number: MFCD00000793
CAS Number: 4124-31-6
EINECS: 223-925-3
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR