Tetrabromomethane, (max. 6% H₂O) 98% (dry weight)

Supplier: Thermo Scientific Chemicals
Warning

L00821-14 L00821-22 L00821-36
CAAAAL00821-14EA 60.2 CAD
CAAAAL00821-14 CAAAAL00821-22 CAAAAL00821-36
Tetrabromomethane, (max. 6% H₂O) 98% (dry weight)
Tetrabromomethane

Purity: 98% CAS No.: 558-13-4 Molecular Formula: CBr4 Formula Weight: 331.63 Melting Point: 90-95° Boiling Point: 190° UN No.: 2516 MDL No.: MFCD00000117

Formula: Br₄C
Boiling Pt: 190 °C
Melting Pt: 90…95 °C
Density: 3.420
Storage Temperature: Ambient
MDL Number: MFCD00000117
CAS Number: 558-13-4
EINECS: 209-189-6
UN: 2516
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR