6-Chloro-m-cresol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00816-14
L00816-22
CAAAAL00816-14EA
90.85
CAD
CAAAAL00816-14
CAAAAL00816-22
6-Chloro-m-cresol 99%
6-Chloro-m-cresol
Purity: 99%
CAS No.: 615-74-7
Molecular Formula: C7H7ClO
Formula Weight: 142.59
Flash Point: 81°(177°F)
Melting Point: 44-48°
Boiling Point: 195-197°
Beilstein Reference: 2041487
UN No.: 3437
MDL No.: MFCD00002175
Formula:
C₇H₇ClO Boiling Pt: 195…197 °C Melting Pt: 44…48 °C Density: 1.215 Flash Pt: 81 °C (177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002175 CAS Number: 615-74-7 EINECS: 210-444-9 UN: 3437 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...