3,5-Dimethoxytoluene 98%

Supplier: Thermo Scientific Chemicals
Warning

L00809-03 L00809-06 L00809-14
CAAAAL00809-03EA 70.31 CAD
CAAAAL00809-03 CAAAAL00809-06 CAAAAL00809-14
3,5-Dimethoxytoluene 98%
3,5-Dimethoxytoluene

Purity: 98% CAS No.: 4179-19-5 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 101°(213°F) Refractive Index: 1.5225 Odor: Aromatic Boiling Point: 244° Beilstein Reference: 2043558 MDL No.: MFCD00015435

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 244 °C
Density: 1,039
Flash Pt: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015435
CAS Number: 4179-19-5
EINECS: 224-048-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR