1-Benzylimidazole 98+%
Supplier: Thermo Scientific Chemicals
L00802-06
L00802-14
CAAAAL00802-06EA
56.41
CAD
CAAAAL00802-06
CAAAAL00802-14
1-Benzylimidazole 98+%
1-Benzylimidazole
Purity: 98+%
CAS No.: 4238-71-5
Molecular Formula: C10H10N2
Formula Weight: 158.20
Melting Point: 68-73°
Boiling Point: 310°
MDL No.: MFCD00005296
Formula:
C₁₀H₁₀N₂ Boiling Pt: 310 °C Melting Pt: 68…73 °C Density: 1.22 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005296 CAS Number: 4238-71-5 EINECS: 224-200-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...