Ethyl indole-2-carboxylate 98+%
Supplier: Thermo Scientific Chemicals
L00797-06
L00797-14
CAAAAL00797-06EA
81.69
CAD
CAAAAL00797-06
CAAAAL00797-14
Ethyl indole-2-carboxylate 98+%
Ethyl indole-2-carboxylate
Purity: 98+%
CAS No.: 3770-50-1
Molecular Formula: C11H11NO2
Formula Weight: 189.21
Melting Point: 122-125°
Beilstein Reference: 146395
Formula:
C₁₁H₁₁NO₂ Melting Pt: 122…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005609 CAS Number: 3770-50-1 EINECS: 223-206-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...