5-Hydroxyindole-2-carboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00786-03
L00786-06
CAAAAL00786-03EA
114.39
CAD
CAAAAL00786-03
CAAAAL00786-06
5-Hydroxyindole-2-carboxylic acid 97%
5-Hydroxyindole-2-carboxylic acid
Purity: 97%
CAS No.: 21598-06-1
Molecular Formula: C9H7NO3
Formula Weight: 177.16
Storage Temperature: 2°C to 8°C
Melting Point: ca 245° dec.
Beilstein Reference: 153214
MDL No.: MFCD00005615
Formula:
C₉H₇NO₃ Melting Pt: ∼ 245 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005615 CAS Number: 21598-06-1 EINECS: 244-468-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...