2,2-Difluorosuccinic acid 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α,α-Difluorosuccinic acid, 2,2-Difluoro-1,4-butanedioic acid

L00773-03 L00773-06
CAAAAL00773-03EA 237 CAD
CAAAAL00773-03 CAAAAL00773-06
2,2-Difluorosuccinic acid 94%
2,2-Difluorosuccinic acid

Purity: 94% CAS No.: 665-31-6 Molecular Formula: C4H4F2O4 Formula Weight: 154.07 Flash Point: 110°(230°F) Melting Point: 139-144° Beilstein Reference: 1773355 UN No.: 3261 MDL No.: MFCD00040970

Formula: C₄H₄F₂O₄
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 139…144 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040970
CAS Number: 665-31-6
EINECS: 211-558-1
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR