3-Buten-1-ol 98+%
Supplier: Thermo Scientific Chemicals
L00767-06
L00767-14
CAAAAL00767-06EA
62.73
CAD
CAAAAL00767-06
CAAAAL00767-14
3-Buten-1-ol 98+%
3-Buten-1-ol
Purity: 98+%
CAS No.: 627-27-0
Molecular Formula: CH2=CHCH2CH2OH
Formula Weight: 72.11
Flash Point: 32°(89°F)
Refractive Index: 1.4210
Boiling Point: 113-114°
Beilstein Reference: 1633504
UN No.: 1987
MDL No.: MFCD00002959
Formula:
C₄H₈O |
MDL Number:
MFCD00002959 CAS Number: 627-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...