4-Nitrodiphenylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00728-03
L00728-06
CAAAAL00728-03EA
92.11
CAD
CAAAAL00728-03
CAAAAL00728-06
4-Nitrodiphenylamine 98+%
4-Nitrodiphenylamine
Purity: 98+%
CAS No.: 836-30-6
Molecular Formula: C12H10N2O2
Formula Weight: 214.22
Flash Point: 190°(374°F)
Melting Point: 132-135°
Boiling Point: 211°/30mm
Beilstein Reference: 2051910
UN No.: 3077
MDL No.: MFCD00007301
Formula:
C₁₂H₁₀N₂O₂ Boiling Pt: 211 °C (30 mmHg) Melting Pt: 132…135 °C Flash Pt: 190 °C (374 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007301 CAS Number: 836-30-6 EINECS: 212-646-2 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...