Bromopyruvic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Bromo-2-oxopropionic acid, 3-Bromopyruvic acid
L00720-04
L00720-09
L00720-18
CAAAAL00720-04EA
62.57
CAD
CAAAAL00720-04
CAAAAL00720-09
CAAAAL00720-18
Bromopyruvic acid 97%
Bromopyruvic acid
Purity: 97%
CAS No.: 1113-59-3
Molecular Formula: C3H3BrO3
Formula Weight: 166.96
Storage Temperature: 2°C to 8°C
Melting Point: 75-80°
Beilstein Reference: 1746786
UN No.: 3261
MDL No.: MFCD00002587
Formula:
BrH₂CC(O)COOH MW: 166.96 g/mol Boiling Pt: 249 Melting Pt: 75…80 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002587 CAS Number: 1113-59-3 EINECS: 214-206-5 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...