4,5-Dimethoxy-2-nitrobenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
6-Nitroveratryl alcohol, (4,5-Dimethoxy-2-nitrophenyl)methanol
L00719-06
L00719-14
CAAAAL00719-06EA
155.95
CAD
CAAAAL00719-06
CAAAAL00719-14
4,5-Dimethoxy-2-nitrobenzyl alcohol 98%
4,5-Dimethoxy-2-nitrobenzyl alcohol
Purity: 98%
CAS No.: 1016-58-6
Molecular Formula: C9H11NO5
Formula Weight: 213.19
Storage Temperature: 2°C to 8°C
Melting Point: 144-148°
Beilstein Reference: 1880093
MDL No.: MFCD00014701
Formula:
C₉H₁₁NO₅ MW: 213.19 g/mol Melting Pt: 144…148 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014701 CAS Number: 1016-58-6 EINECS: 213-806-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...