(2-Hydroxyethyl)triphenylphosphonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

L00679-14 L00679-22
CAAAAL00679-14EA 167.48 CAD
CAAAAL00679-14 CAAAAL00679-22
(2-Hydroxyethyl)triphenylphosphonium bromide 98+%
(2-Hydroxyethyl)triphenylphosphonium bromide

Purity: 98+% CAS No.: 7237-34-5 Molecular Formula: HO(CH2)2P(C6H5)3Br Formula Weight: 387.26 Melting Point: 218-220° Beilstein Reference: 3922741

Formula: C₂₀H₂₀BrOP
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00011901
CAS Number: 7237-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR