Triethylsulfonium iodide 97%

Supplier: Thermo Scientific Chemicals
Warning

L00667-06 L00667-14 L00667-22
CAAAAL00667-06EA 75.05 CAD
CAAAAL00667-06 CAAAAL00667-14 CAAAAL00667-22
Triethylsulfonium iodide 97%
Triethylsulphonium iodide

Purity: 97% CAS No.: 1829-92-1 Molecular Formula: (CH3CH2)3SI Formula Weight: 246.15 Storage Temperature: 2°C to 8°C Melting Point: ca 148° dec. Beilstein Reference: 3911790 MDL No.: MFCD00051867

Formula: C₆H₁₅IS
Melting Pt: ∼ 148 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051867
CAS Number: 1829-92-1
EINECS: 217-383-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR