3,5-Dibromo-p-cresol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00660-03
L00660-06
CAAAAL00660-03EA
129.56
CAD
CAAAAL00660-03
CAAAAL00660-06
3,5-Dibromo-p-cresol 97%
3,5-Dibromo-p-cresol
Purity: 97%
CAS No.: 13979-81-2
Molecular Formula: C7H6Br2O
Formula Weight: 265.94
Melting Point: 106-110°
Beilstein Reference: 2249692
Formula:
C₇H₆Br₂O Melting Pt: 106…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013968 CAS Number: 13979-81-2 EINECS: 237-763-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...