N,N-Dimethylformamide dipropyl acetal 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,1-Dipropoxytrimethylamine, N,n-dimethylformamidedipropylacetal, 1,1-Dipropoxy-N,N-dimethylmethylamine
L00643-09
L00643-18
CAAAAL00643-09EA
86.27
CAD
CAAAAL00643-09
CAAAAL00643-18
N,N-Dimethylformamide dipropyl acetal 97%
N,N-Dimethylformamide dipropyl acetal
Purity: 97%
CAS No.: 6006-65-1
Molecular Formula: C9H21NO2
Formula Weight: 175.27
Flash Point: 46°(115°F)
Refractive Index: 1.4088
Boiling Point: 67-68°/14mm
Beilstein Reference: 1098524
UN No.: 1993
MDL No.: MFCD00009374
Formula:
C₉H₂₁NO₂ MW: 175.27 g/mol Boiling Pt: 67…68 °C/14 mm Density: 0.854 Flash Pt: 46 °C (115 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009374 CAS Number: 6006-65-1 EINECS: 227-855-4 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...