Ethyl-O-toluate 98+%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Ethyl-2-methylbenzoate, 2-Toluic acid ethyl ester, o-Toluic acid ethyl ester, Ethyl 2-methylbenzoate, 2-Methylbenzoic acid ethyl ester
L00598-14
L00598-22
CAAAAL00598-14EA
75.37
CAD
CAAAAL00598-14
CAAAAL00598-22
Ethyl-O-toluate 98+%
Ethyl o-toluate
Purity: 98+%
CAS No.: 87-24-1
Molecular Formula: C10H12O2
Formula Weight: 164.21
Flash Point: 91°(195°F)
Refractive Index: 1.5070
Boiling Point: 227°
Beilstein Reference: 2042517
MDL No.: MFCD00009112
Formula:
C₁₀H₁₂O₂ MW: 164.2 Boiling Pt: 227 Melting Pt: -10 Density: 1.033 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009112 CAS Number: 87-24-1 EINECS: 201-734-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...