Ethyl bromopyruvate ≥75%, tech.

Supplier: Thermo Scientific Chemicals
Danger

L00582-06 L00582-14 L00582-22
CAAAAL00582-06EA 47.56 CAD
CAAAAL00582-06 CAAAAL00582-14 CAAAAL00582-22
Ethyl bromopyruvate ≥75%, tech.
Ethyl bromopyruvate
Formula: BrH₂CC(O)COOCH₂CH₃
Boiling Pt: 98…100 °C/10 mm
Density: 1.555
Flash Pt: 98 °C (208 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000204
CAS Number: 70-23-5
EINECS: 200-729-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR