N,N,N',N'-Tetramethyl-p-phenylenediamine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N,N,N,N-Tetramethyl-1,4-phenylenediamine, N,N,N′,N′-Tetramethyl-p-phenylenediamine, TMPD, N,N,N',N'-Tetramethyl-1,4-phenylenediamine
L00559-09
L00559-18
CAAAAL00559-09EA
98.12
CAD
CAAAAL00559-09
CAAAAL00559-18
N,N,N',N'-Tetramethyl-p-phenylenediamine 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine
Purity: 98+%
CAS No.: 100-22-1
Molecular Formula: C10H16N2
Formula Weight: 164.25
Storage Temperature: 2°C to 8°C
Melting Point: 48-52°
Boiling Point: 260°
Beilstein Reference: 1564025
Merck Reference: 14,9228
UN No.: 1673
MDL No.: MFCD00008309
Formula:
C₁₀H₁₆N₂ MW: 164.25 g/mol Boiling Pt: 260 °C Melting Pt: 48…52 °C Density: 1.08 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008309 CAS Number: 100-22-1 EINECS: 202-831-6 UN: 2811 ADR: 6.1,III Merck Index: 14,9228 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...