Tropone 97%

Supplier: Thermo Scientific Chemicals

L00473-03 L00473-06
CAAAAL00473-03EA 113.76 CAD
CAAAAL00473-03 CAAAAL00473-06
Tropone 97%
Tropone

Purity: 97% CAS No.: 539-80-0 Molecular Formula: C7H6O Formula Weight: 106.12 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.6150 Boiling Point: 91-92°/4mm Beilstein Reference: 1902335 MDL No.: MFCD00014331

Formula: C₇H₆O
Boiling Pt: 91…92 °C/4 mm
Density: 1.094
Flash Pt: >110 °C(230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00014331
CAS Number: 539-80-0
EINECS: 208-725-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR