Hexaphenylbenzene 98+%
Supplier: Thermo Scientific Chemicals
L00446-03
L00446-06
CAAAAL00446-03EA
116.6
CAD
CAAAAL00446-03
CAAAAL00446-06
Hexaphenylbenzene 98+%
Hexaphenylbenzene
Purity: 98+%
CAS No.: 992-04-1
Molecular Formula: C42H30
Formula Weight: 534.70
Melting Point:
Formula:
C₄₂H₃₀ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003057 CAS Number: 992-04-1 EINECS: 213-591-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...