3'-Methylacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00440-06
L00440-14
CAAAAL00440-06EA
46.93
CAD
CAAAAL00440-06
CAAAAL00440-14
3'-Methylacetophenone 97%
3'-Methylacetophenone
Purity: 97%
CAS No.: 585-74-0
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 85°(185°F)
Refractive Index: 1.5310
Melting Point: -9°
Boiling Point: 219°
Beilstein Reference: 956637
MDL No.: MFCD00008742
Formula:
C₉H₁₀O Boiling Pt: 219 °C Melting Pt: –9 °C Density: 1.010 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008742 CAS Number: 585-74-0 EINECS: 209-561-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...