N,N-Dimethylformamide di-tert-butyl acetal tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
L00431-06
L00431-14
CAAAAL00431-06EA
178.54
CAD
CAAAAL00431-06
CAAAAL00431-14
N,N-Dimethylformamide di-tert-butyl acetal tech. 90%, Technical Grade
N,N-Dimethylformamide di-tert-butyl acetal
Purity: 90%
CAS No.: 36805-97-7
Molecular Formula: C11H25NO2
Formula Weight: 203.33
Flash Point: 33°(91°F)
Refractive Index: 1.4120
Boiling Point: 75-77°/12mm
Beilstein Reference: 969629
UN No.: 1993
MDL No.: MFCD00015002
Formula:
C₁₁H₂₅NO₂ Boiling Pt: 75…77 °C/12 mm Density: 0.850 Flash Pt: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015002 CAS Number: 36805-97-7 EINECS: 253-222-7 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...