N,N-Dimethyl-4-nitroaniline 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L00404-06
L00404-14
CAAAAL00404-06EA
68.26
CAD
CAAAAL00404-06
CAAAAL00404-14
N,N-Dimethyl-4-nitroaniline 98+%
N,N-Dimethyl-4-nitroaniline
Purity: 98+%
CAS No.: 100-23-2
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Melting Point: 163-166°
Beilstein Reference: 638087
UN No.: 2811
MDL No.: MFCD00014712
Formula:
C₈H₁₀N₂O₂ MW: 166.18 g/mol Boiling Pt: 288 °C (1013 hPa) Melting Pt: 163…166 °C Density: 1.179 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014712 CAS Number: 100-23-2 EINECS: 202-832-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...