Ethyl diethoxyacetate 97+%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Diethoxyacetic ethyl ester
L00390-14
L00390-22
CAAAAL00390-14EA
116.45
CAD
CAAAAL00390-14
CAAAAL00390-22
CAAAAL00390-36
Ethyl diethoxyacetate 97+%
Ethyl diethoxyacetate
Purity: 97+%
CAS No.: 6065-82-3
Molecular Formula: C8H16O4
Formula Weight: 176.21
Flash Point: 85°(185°F)
Refractive Index: 1.4100
Boiling Point: 198-200°
Beilstein Reference: 878579
MDL No.: MFCD00009231
Formula:
C₈H₁₆O₄ MW: 176.21 g/mol Boiling Pt: 198…200 °C Density: 0.988 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009231 CAS Number: 6065-82-3 EINECS: 227-999-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...